Complete Meter Stick Optics Bench
Complete Meter Stick Optics Bench, 1
Catalog # 470219-700
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: OBSET1
Jump to:
Product Details & Documents
Classical experiment for studying optics on a budget.
- All components connect to any standard meter stick
Student optical bench contains sufficient equipment for high school physics applications.
Contents:
1 Meter Stick
1 Screen support
2 lens supports for 38mm and 75mm lenses/mirrors
5 Board screens measuring 10 x 125 cm with a millimeter scale on both sides
1 Marker and object
2 Metal supports for meter stick
1 Candle holder
1 Paraffin candle
1 Activity guide
Required but not included are lenses. Focal lengths under 50cm work best.
Documents
Especificaciones
- Description:Complete Meter Stick Optics Bench
- No. of Lab Groups:1