Orders LinkContactUsLinkComponent
PREempt™ Concentrate Disinfectant, Contec®
PREempt™ Concentrate Disinfectant, Contec®
PREempt™ Concentrate, 200 L (55 gal.)
Catalog # 10822-522
PREempt™ Concentrate Disinfectant, Contec®
Catalog # 10822-522
Supplier:  
PREempt™ Concentrate Disinfectant, Contec®
PREempt™ Concentrate Disinfectant, Contec®
PREempt™ Concentrate, 200 L (55 gal.)
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
PREEMPT CONCENTRATE 1G DISINFECTANT CS4 Catalog #10822-520
Product 10822-520 is similar to 10822-522 but manufacturer and specifications may vary.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PREempt™ Concentrate Disinfectant Solution works fast for large areas to ensure user, protocol, and product compliance with fast contact times and broad-spectrum efficacy
  • 3 minute Broad-Spectrum Sanitizing (1:128)
  • 30 second Broad-Spectrum Sanitizing (1:16)
PREempt™ Concentrate utilizes AHP®, a patented synergistic blend of commonly used and safe ingredients which, when combined with low levels of hydrogen peroxide, dramatically increase the potency and cleaning performance.
The concentrated formula requires dilution and is cost effective for large surfaces.
Specifications
  • Description:
    PREempt™ Concentrate
  • Size:
    200 L (55 gal.)