CENCO® Spectrometer Prisms
Prism Borosilicate Crown Glass, 1.51
Catalog # 470219-498
Supplier: EISCO SCIENTIFIC LLC
: PH0583A
Jump to:
Product Details & Documents
Spectrometer prisms offer a different index of refraction for every need.
- Different Indices of Refractions for Every Need
- Glass Material
- Designed to Yield Accurate Results
Although any prism can be used in a spectrometer, these are specially designed to yield accurate results for spectroscopy experiments.
Choose your prism type and the approximate index of refraction that would work best for your experiment.
Especificaciones
- Description:Prism Borosilicate Crown Glass, 1.51