Optical Prisms
Acrylic, 55 mm, 38 mm
Catalog # 470219-460
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: PH0562B
Jump to:
Product Details & Documents
Right angle and equilateral prisms are available in a variety of forms.
- For Investigating Refraction and Creating Rainbows
- Non-Optically Worked
- Polished Faces
- Beveled Edges
- Right Angle and Equilateral Prisms Available
These prisms are non-optically worked with polished faces and edges slightly beveled. They're useful for investigating refraction and creating rainbows.
Choose the size and material that best fits your experimental needs.
Especificaciones
- Material:Acrylic
- Size:55 mm, 38 mm