Orders LinkContactUsLinkComponent
Optical Prisms
Glass, 25 mm
  470219-446
 :  PH0558A
Optical Prisms
  470219-446
Optical Prisms
Optical Prisms
Glass, 25 mm
$3.50
  31/12/2026
Each (1UN)
Right angle and equilateral prisms are available in a variety of forms.
  • For Investigating Refraction and Creating Rainbows
  • Non-Optically Worked
  • Polished Faces
  • Beveled Edges
  • Right Angle and Equilateral Prisms Available
These prisms are non-optically worked with polished faces and edges slightly beveled. They're useful for investigating refraction and creating rainbows.
Choose the size and material that best fits your experimental needs.
  • Material:
    Glass
  • Size:
    25 mm