Orders LinkContactUsLinkComponent
CENCO® Spectrometer Prisms
CENCO® Spectrometer Prisms
Prism Flint Glass, 1.65
Catalog # 470219-502
Supplier #:  PH0583C
CENCO® Spectrometer Prisms
Catalog # 470219-502
CENCO® Spectrometer Prisms
CENCO® Spectrometer Prisms
Prism Flint Glass, 1.65
Price
$1,531.89
Log in for availability
Each (1UN)
Product Details & Documents
Spectrometer prisms offer a different index of refraction for every need.
  • Different Indices of Refractions for Every Need
  • Glass Material
  • Designed to Yield Accurate Results
Although any prism can be used in a spectrometer, these are specially designed to yield accurate results for spectroscopy experiments.
Choose your prism type and the approximate index of refraction that would work best for your experiment.
Specifications
  • Description:
    Prism Flint Glass, 1.65