Optical Refraction Components
Semi Circular Block, Acrylic, 90 x 16 mm
470219-484
: PH0570A
These Optical Refraction Components are made from imported optical glass or acrylic with optically worked surfaces.
- Students Measure the Refraction of Light
- Refraction Blocks, Prisms, and Lenses
- Optically Worked Surfaces
- Bubble-Free Glass
- Ideal in Conjunction with a Light Box
All angles are accurately worked with edges slightly beveled and glass free from bubbles.
These optical elements are perfect to use with a light box, so students can study and measure the refraction of light through different mediums and different shapes.
- Description:Semi Circular Block
- Dimensions:90 x 16 mm
- Material:Acrylic