Orders LinkContactUsLinkComponent
Optical Prisms
Optical Prisms
Acrylic, 74 mm, 50 mm
Catalog # 470219-462
Supplier #:  PH0562C
Optical Prisms
Catalog # 470219-462
Optical Prisms
Optical Prisms
Acrylic, 74 mm, 50 mm
Price
$357.65
Log in for availability
Each (1UN)
Product Details & Documents
Right angle and equilateral prisms are available in a variety of forms.
  • For Investigating Refraction and Creating Rainbows
  • Non-Optically Worked
  • Polished Faces
  • Beveled Edges
  • Right Angle and Equilateral Prisms Available
These prisms are non-optically worked with polished faces and edges slightly beveled. They're useful for investigating refraction and creating rainbows.
Choose the size and material that best fits your experimental needs.
Specifications
  • Material:
    Acrylic
  • Size:
    74 mm, 50 mm