Orders LinkContactUsLinkComponent
Optical Prisms
Acrylic, 35 mm, 25 mm
  470219-458
 :  PH0562A
Optical Prisms
  470219-458
Optical Prisms
Optical Prisms
Acrylic, 35 mm, 25 mm
$133.26
Each (1UN)
Right angle and equilateral prisms are available in a variety of forms.
  • For Investigating Refraction and Creating Rainbows
  • Non-Optically Worked
  • Polished Faces
  • Beveled Edges
  • Right Angle and Equilateral Prisms Available
These prisms are non-optically worked with polished faces and edges slightly beveled. They're useful for investigating refraction and creating rainbows.
Choose the size and material that best fits your experimental needs.
  • Material:
    Acrylic
  • Size:
    35 mm, 25 mm