Orders LinkContactUsLinkComponent
Grindometers, Industrial Physics
Grindometers, Industrial Physics
Grindometer, Double Groove, 0 - 15 µm, Graduation 1.5 µm
Catalog # 10025-134
Número de proveedor #:  VF2110
Grindometers, Industrial Physics
Catalog # 10025-134
Grindometers, Industrial Physics
Grindometers, Industrial Physics
Grindometer, Double Groove, 0 - 15 µm, Graduation 1.5 µm
Price
$28,339.50
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
These grindometers or gauges of fineness are precision instruments to determine particle size and fineness of many materials like paints, lacquers, pigments, filler, chocolate etc
  • Precision instrument
  • Ergonomic shaped scraper for an easy grip.
  • Many models available
Gauge and bevelled scraper are made of hardened medical grade stainless tool steel and have an accuracy of 2μm.
TQC Grindometers have double grooves with graded slopes graduated in three different parameters: µm (microns), NS (Hegman) and PCU (North).
Certifications: ASTM D1210, ASTM D1316, DIN 53203, DIN EN ISO NF21524, FTMS 141 a M.4411.1 ISO1524, NFT 30 046.
Delivery information: All models come with a base and a scraper.
Especificaciones
  • Description:
    Grindometer, Double Groove, 0 - 15 µm, Graduation 1.5 µm