Order Entry
Mexico
Orders LinkContactUsLinkComponent
General Purpose Viscosity Standards, Cannon Instruments

General Purpose Viscosity Standards, Cannon Instruments

Viscosity standard N750, 500 ml, (20°C) 2200 mPa·s
Catalog # 10021-270
Número de proveedor #:  9727-C51.016
General Purpose Viscosity Standards, Cannon Instruments
Catalog # 10021-270
CAS Number:  
General Purpose Viscosity Standards, Cannon Instruments

Viscosity standard N750, 500 ml, (20°C) 2200 mPa·s

CAS Number:  
Price
$3,961.03
Log in Para disponibilidad
Each (1ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These certified standards are useful for calibration and verification procedures.

  • It is suitable for all types of viscometers
  • It also helps ensures the reliability of measurements

Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.

Documents

Especificaciones

Especificaciones

  • Description:
    Viscosity standard N750
  • Size:
    500 ml
  • Viscosity:
    (20°C) 2200 mPa·s

Especificaciones

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.