Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

General Purpose Viscosity Standards, Cannon Instruments

Viscosity standard S200, 500 ml, (20°C) 460 mPa·s
Catalog # 10021-224
 :  9727-C45.016
General Purpose Viscosity Standards, Cannon Instruments
Catalog # 10021-224
CAS Number:  
General Purpose Viscosity Standards, Cannon Instruments

Viscosity standard S200, 500 ml, (20°C) 460 mPa·s

CAS Number:  
Price
$3,520.88
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These certified standards are useful for calibration and verification procedures.

  • It is suitable for all types of viscometers
  • It also helps ensures the reliability of measurements

 : Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.

 

Specifications

Specifications

  • Description:
    Viscosity standard S200
  • Size:
    500 ml
  • Viscosity:
    (20°C) 460 mPa·s

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.