Orders LinkContactUsLinkComponent
General Purpose Viscosity Standards, Cannon Instruments
Viscosity Standard S20, 500 ml, (20°C) 37 mPa·s
  10021-170
 :  9727-C35.016
General Purpose Viscosity Standards, Cannon Instruments
  10021-170
 :  
General Purpose Viscosity Standards, Cannon Instruments
Viscosity Standard S20, 500 ml, (20°C) 37 mPa·s
 :  
$3,520.88
Each (1ml)
These certified standards are useful for calibration and verification procedures.
  • It is suitable for all types of viscometers
  • It also helps ensures the reliability of measurements
Certifications: Meets the precision specifications of ASTM D 445/446 and ISO 3104/3105, and are traceable to the NIST.
Documents
  • Description:
    Viscosity Standard S20
  • Size:
    500 ml
  • Viscosity:
    (20°C) 37 mPa·s
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 