Orders LinkContactUsLinkComponent
Ward's® Apatite (Massive)
Ward's® Apatite (Massive)
Apatite (massive), bulk pack, 1 kg, Size will vary
Catalog # 470218-158
Supplier:   Avantor
Ward's® Apatite (Massive)
Catalog # 470218-158
Supplier:  Avantor
Ward's® Apatite (Massive)
Ward's® Apatite (Massive)
Apatite (massive), bulk pack, 1 kg, Size will vary
Price
$747.43
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Blue fluorapatite, pure crystalline, Moh's hardness of 5.
  • Collected in Madagascar
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25 to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Especificaciones
  • Description:
    Apatite (massive), bulk pack, 1 kg
  • Specimen Dimensions:
    Size will vary