Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

1,2-Dichloroethane ≥99%

500 ml
Catalog # BT223375-500ML
 :  223375-500ML
1,2-Dichloroethane ≥99%
Catalog # BT223375-500ML
CAS Number:  107-06-2
1,2-Dichloroethane ≥99%
1,2-Dichloroethane ≥99%

500 ml

CAS Number:  107-06-2
Price
$550.00
Log in Para disponibilidad
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS: 107-06-2; EC No: 203-458-1; MDL No: MFCD00000963; RTECS: KI0525000 UN No: UN1184; Haz Class: 3 (6.1); Packing Group: II Liquid; Linear Formula: ClCH2CH2Cl ; Molecular Formula: C2H4Cl2; MW: 98.96 Melting Point: -35°; Boiling Point: 83°; Flash point: 13°C (55°F) Density (g/mL): 1.256; Refractive Index: 1.444

  • RTECS: KI0525000
  • Refractive index: 1.444
  • Liquid
Sinónimos:  Ethylene chloride, Ethylene dichloride, Dichloroethane
  • CAS Number:107-06-2
  • Fórmula:ClH₂CCH₂Cl
  • Número MDL:MFCD00000963
  • Molecular Weight:98.96 g/mol
  • Punto de ebullición:83 °C
  • Punto de fusión:–35 °C
  • Densidad:1.256 g/mL
  • Punto de inflamación:13 °C (55 °F)
  • EINECS:203-458-1
  • UN:1184
  • ADR:3,II
Especificaciones

Resultados de prueba de especificación

  • Assay:
    99%

Especificaciones

  • Size:
    500 ml

Especificaciones

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.