Triangular Base and Rod Assembly
230 mm, 60 cm
Catalog # 470165-334
Supplier: EISCO SCIENTIFIC LLC
: TRISTD2
Jump to:
Product Details & Documents
Sturdy assembly for all of your laboratory needs.
- Heavy-duty cast iron construction
- Two sizes available
Triangular-shaped, cast iron base provides stability. Base is 23mm wide (radius: 13cm). Each size includes a threaded steal rod.
Specifications
- Rod Length:60 cm
- Base Dimensions:230 mm