Order Entry
Mexico
Orders LinkContactUsLinkComponent
A-Shaped Base Retort Stand

A-Shaped Base Retort Stand

11" Base with Stainless Steel Rod, 11 mm, 750 mm, 10 x 1.5 mm
  470165-372
Supplier #:  STDRDAB03
A-Shaped Base Retort Stand
  470165-372
A-Shaped Base Retort Stand
A-Shaped Base Retort Stand

11" Base with Stainless Steel Rod, 11 mm, 750 mm, 10 x 1.5 mm

 
$1,338.91
   
Each (1UN)
 

This cast iron, powder-coated, A-shaped base includes a stainless steel rod.

  • Powder-Coated
  • Heavy-Duty
  • A-Shaped
  • Multiple Sizes Available
  • Stainless Steel Rod Included

These bases can be nested in one another.

Multiple sizes are available.

 
  • Rod Length:
    750 mm
  • Description:
    11" Base with Stainless Steel Rod
  • Rod Diameter:
    11 mm
  • Thread Size:
    10 x 1.5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.