Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

A-Shaped Base Retort Stand

9" Base with Stainless Steel Rod, 11 mm, 600 mm, 10 x 1.5 mm
  470165-370
 :  STDRDAB02
A-Shaped Base Retort Stand
  470165-370
A-Shaped Base Retort Stand
A-Shaped Base Retort Stand

9" Base with Stainless Steel Rod, 11 mm, 600 mm, 10 x 1.5 mm

 
$1,127.32
   
Each (1UN)
 

This cast iron, powder-coated, A-shaped base includes a stainless steel rod.

  • Powder-Coated
  • Heavy-Duty
  • A-Shaped
  • Multiple Sizes Available
  • Stainless Steel Rod Included

These bases can be nested in one another.

Multiple sizes are available.

 
  • Rod Length:
    600 mm
  • Description:
    9" Base with Stainless Steel Rod
  • Rod Diameter:
    11 mm
  • Thread Size:
    10 x 1.5 mm