Tripod Triangle Stands
Tripod Triangle Stand for burners, 21 cm, 15 cm
Catalog # 470165-366
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: CH0645B
Jump to:
Product Details & Documents
Zinc plated stands feature cast iron top and mild steel legs. Multiple sizes available.
Especificaciones
- Ext.Ø:15 cm
- Description:Tripod Triangle Stand for burners
- Height:21 cm



