Orders LinkContactUsLinkComponent
Celestron UpClose Binoculars
Celestron UpClose Binoculars
UpClose G2 Zoom Porro Binocular
Catalog # 470164-702
Supplier #:  71260
Celestron UpClose Binoculars
Catalog # 470164-702
Celestron UpClose Binoculars
Celestron UpClose Binoculars
UpClose G2 Zoom Porro Binocular
Price
$2,365.50
Log in for availability
Each (1UN)
Product Details & Documents
High-quality binoculars at an affordable price.
  • Multi-coated optics increase light transmission resulting in brighter and sharper images with high contrast levels
  • Ergonomic thumb indents and finger ridges for extra non-slip comfort and ease of use
  • Water-resistant
  • Aligned with GLOBE protocols
These binoculars are perfect for a variety of activities, such as bird watching, sporting events, nature and wildlife, hunting, travel and ventures, concerts and live events, and boating.
Delivery information: Binocular straps and soft carrying case included.
Specifications
  • Magnification:
    10-30×
  • Description:
    UpClose G2 Zoom Porro Binocular