Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Pull-Tite™ Drawstring Storage Bags, Elkay Plastics

Double drawstring bags, 2 mil., 6×8", Clear
Catalog # 10790-222
 :  DS20608
Pull-Tite™ Drawstring Storage Bags, Elkay Plastics
Catalog # 10790-222
Pull-Tite™ Drawstring Storage Bags, Elkay Plastics
Pull-Tite™ Drawstring Storage Bags, Elkay Plastics

Double drawstring bags, 2 mil., 6×8", Clear

Price
$4,086.39
Log in Para disponibilidad
Case (1,000UN)
Product Details & Documents

A double pull drawstring opens easily and provides a secure closure and a means of hanging the bag.

  • Available in both clear and clear with a white write-on block
  • Easy product labeling and identification

Some of the uses include: parts, work orders, manuals, samples, personal belongings and more.

Certifications: This product meets FDA requirements for use in food applications.

Ordering information: Bags 15" wide and under come in dispenser packs of 100.

Especificaciones
  • Color:
    Clear
  • Description:
    Double drawstring bags
  • Dimensions:
    6×8"
  • Thickness:
    2 mil.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.