Order Entry
Mexico
Orders LinkContactUsLinkComponent
Pull-Tite™ Drawstring Storage Bags, Elkay Plastics

Pull-Tite™ Drawstring Storage Bags, Elkay Plastics

Double drawstring bags, 1.5 mil., 7×12", Clear
Catalog # 10789-874
Supplier #:  DS15712
Pull-Tite™ Drawstring Storage Bags, Elkay Plastics
Catalog # 10789-874
Pull-Tite™ Drawstring Storage Bags, Elkay Plastics
Pull-Tite™ Drawstring Storage Bags, Elkay Plastics

Double drawstring bags, 1.5 mil., 7×12", Clear

Price
$4,607.64
Log in for availability
Case (1,000UN)
Product Details & Documents

A double pull drawstring opens easily and provides a secure closure and a means of hanging the bag.

  • Available in both clear and clear with a white write-on block
  • Easy product labeling and identification

Some of the uses include: parts, work orders, manuals, samples, personal belongings and more.

 : This product meets FDA requirements for use in food applications.

 : Bags 15" wide and under come in dispenser packs of 100.

Specifications
  • Color:
    Clear
  • Description:
    Double drawstring bags
  • Dimensions:
    7×12"
  • Thickness:
    1.5 mil.