Orders LinkContactUsLinkComponent
Examination Gloves, Microflex®, Ansell
Examination Gloves, Microflex®, Ansell
S, Blue, 24 cm (9⅜")
Catalog # 10770-436
Supplier:   Ansell Healthcare
Supplier #:  N211
Examination Gloves, Microflex®, Ansell
Catalog # 10770-436
Supplier:  Ansell Healthcare
Examination Gloves, Microflex®, Ansell
Examination Gloves, Microflex®, Ansell
S, Blue, 24 cm (9⅜")
Price
$347.00
Log in for availability
Paquete de 100 Case de 10 (100UN)
Product Details & Documents
This thicker version of the popular Microflex® Cobalt® X N21 glove offers fit and feel with the extra protection of a thicker glove.
  • Fully textured to provide grip in any condition
  • Non sterile
  • Ambidextrous
  • Gloves are blue and feature beaded cuffs
For use in life science, electronics, and food processing industries for analytical testing/measurements; assembly and inspection; biotechnologies; cleaning; food processing; handling small parts; laboratory analysis; laboratory testing; light duty maintenance and clean up; light assembly tasks; maintaining equipment and instruments; pharmaceutical manufacturing; preparing pharmaceutical products; sanitation and dish washing; and as an under glove for meat/fish processing.
Specifications
  • Glove Color:
    Blue
  • Glove size:
    S
  • Length:
    24 cm (9⅜")
  • Palm thickness:
    4.7 mil
  • Cat. No.:
    10770-436
  • Cuff thickness:
    3.9 mil
  • Cuff style:
    Beaded
  • Ambidextrous / hand-specific:
    Ambidextrous
  • Finger thickness:
    5.5 mil