Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Crystallizing Dishes

VWR® Crystallizing Dishes

1800 ml, 170 mm, 90 mm
Catalog # 10754-780
Supplier:   VWR International
VWR® Crystallizing Dishes
Catalog # 10754-780
Supplier:  VWR International
VWR® Crystallizing Dishes
VWR® Crystallizing Dishes

1800 ml, 170 mm, 90 mm

Price
$2,462.59
Log in Para disponibilidad
Paquete de 4 Case de 2 (4UN)
Product Details & Documents

Made from high quality Type 1 Class A, 33 expansion Borosilicate glass, these crystallizing dishes have straight sides and flat bottom.

Documents

Especificaciones
  • Ø:
    170 mm
  • Capacity:
    1800 ml
  • Height:
    90 mm