Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Crystallizing Dishes

VWR® Crystallizing Dishes

700 ml, 125 mm, 65 mm
Catalog # 10754-776
Supplier:   VWR International
VWR® Crystallizing Dishes
Catalog # 10754-776
Supplier:  VWR International
VWR® Crystallizing Dishes
VWR® Crystallizing Dishes

700 ml, 125 mm, 65 mm

Price
$1,464.00
Log in Para disponibilidad
Paquete de 4 Case de 3 (4UN)
Product Details & Documents

Made from high quality Type 1 Class A, 33 expansion Borosilicate glass, these crystallizing dishes have straight sides and flat bottom.

Documents

Especificaciones
  • Ø:
    125 mm
  • Capacity:
    700 ml
  • Height:
    65 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.