VWR® Crystallizing Dishes
80 ml, 60 mm, 35 mm
Supplier: VWR International
Jump to:
Product Details & Documents
Made from high quality Type 1 Class A, 33 expansion Borosilicate glass, these crystallizing dishes have straight sides and flat bottom.
Documents
Especificaciones
- Ø:60 mm
- Capacity:80 ml
- Height:35 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



