3 Wire Sonometer
Product Details & Documents
This 3-Wire Sonometer is a perfect way to illustrate the resonance of a wire based on tension and wire diameter.
- Scaled Divisions
- Three Wires Included
- Three Movable Bridges Included
- Wire Length: 1.5 m
- Wires Made of Rust-Resistant Steel Wires
A spring balance reading to 5N x 100N is fitted to one wire, and the tension exerted and measured can be adjusted by a wing-nut. Another wire passes over a pulley and can be loaded as desired. A third wire is provided for comparison. Its tension may be adjusted by a wrest pin.
The wires are made of steel to avoid rust. The distance between the fixed bridges is 720 mm.
Accessories information: Replacement steel wires are available separately.
Ordering information: This kit includes three movable bridges, a wrest-pin key and a set of four 1.5 m long wires. The SWG sizes are 20, 22, 24, and 26 without weights. A hooked mass set is required but not included.
Documents
Manual 3 Wire Sonometer
Manual
Especificaciones
- Description:Replacement Wire, Set of 6
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



