Accessories for Solvent Pump, Bürkle
Anti-Static Set, When decanting flammable liquids without the machines and containers involved in the process being sufficiently grounded, electrostatic build-up can occur, which for many substances can result in fire or an explosion; Burkle’s anti-static set ensures easy and reliable grounding; the anti-static set consists of three color-coded connecting copper cables with clamps in a handy plastic case
Catalog # 10217-236
Supplier: BURKLE INC
Supplier #: 5602-1000
Jump to:
Product Details & Documents
Specifications
- Description:Anti-Static Set
- For Use With:When decanting flammable liquids without the machines and containers involved in the process being sufficiently grounded, electrostatic build-up can occur, which for many substances can result in fire or an explosion; Burkle’s anti-static set ensures easy and reliable grounding; the anti-static set consists of three color-coded connecting copper cables with clamps in a handy plastic case