Orders LinkContactUsLinkComponent
Accessories for Solvent Pump, Bürkle
Accessories for Solvent Pump, Bürkle
Anti-Static Set, When decanting flammable liquids without the machines and containers involved in the process being sufficiently grounded, electrostatic build-up can occur, which for many substances can result in fire or an explosion; Burkle’s anti-static set ensures easy and reliable grounding; the anti-static set consists of three color-coded connecting copper cables with clamps in a handy plastic case
Catalog # 10217-236
Supplier:   BURKLE INC
Supplier #:  5602-1000
Accessories for Solvent Pump, Bürkle
Catalog # 10217-236
Supplier:  BURKLE INC
Accessories for Solvent Pump, Bürkle
Accessories for Solvent Pump, Bürkle
Anti-Static Set, When decanting flammable liquids without the machines and containers involved in the process being sufficiently grounded, electrostatic build-up can occur, which for many substances can result in fire or an explosion; Burkle’s anti-static set ensures easy and reliable grounding; the anti-static set consists of three color-coded connecting copper cables with clamps in a handy plastic case
Price
$2,854.48
Log in for availability
Each (1UN)
Product Details & Documents
Specifications
  • Description:
    Anti-Static Set
  • For Use With:
    When decanting flammable liquids without the machines and containers involved in the process being sufficiently grounded, electrostatic build-up can occur, which for many substances can result in fire or an explosion; Burkle’s anti-static set ensures easy and reliable grounding; the anti-static set consists of three color-coded connecting copper cables with clamps in a handy plastic case