OTAL Hand Pumps, Bürkle
OTAL Hand pump, rigid Polypropylene delivery tube, reusable, 10 mm (0.39") tube Ø, 4 L/min. (0.14 cfm), 450 mm (17.7") Depth
Catalog # 10216-952
Supplier: BURKLE INC
Supplier #: 5005-1000
Jump to:
Product Details & Documents
Delivery tubes made of PP are resistant against weak acids, caustic solutions and detergents.
Delivery tubes made of PVDF are especially for acids and caustic solutions; not for strongly oxidizing acids and caustic solutions.
Delivery tubes made of stainless steel 304 are for solvents and liquids of VbF Hazard Class AIII (not easily flammable).
Battery pump for safe filling of car batteries with acid or distilled water.
- Compact, space saving pumps
- With universal bungs for container openings of 30-70 mm dia.
- The OTAL mini pump made of PP (model 5005-1000) has a universal bung for container openings of 20-36 mm (0.79-1.42") dia.
Specifications
- Flow rate:4 L/min. (0.14 cfm)
- Description:OTAL Hand pump, rigid Polypropylene delivery tube, reusable, 10 mm (0.39") tube Ø
- Immersion:450 mm (17.7") Depth
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



