Freeze-Thaw Chambers
Product Details & Documents
For precise environmental control, modelling short-term low temperature conditions. This broad range encompasses nearly every ambient environment on earth. Automatic, programmable, and accurate cycling between setpoints enables continuous simulation of harsh freezing and thawing conditions for rapid and repeatable materials testing.
- Wide −25 to +70 °C temperature range for full simulation of freeze/thaw cycles and temperature excursions
- Easy cycle programming through large-format color touchscreen
- Feature up to 5 user-defined programs, with 50 individual steps, up to 255 loops, and a ramp based on any time or rate
- Available humidity control
Caron's chambers are built for many types of freeze/thaw studies, including pharmaceutical drug excursion testing, concrete and asphalt testing, coating and adhesive testing, outdoor weathering tests, and food, beverage, and packaging testing.
Multiple optional accessories are also available to allow you to customize the chamber to match your specific needs.
Certifications: CE.
Ordering information: Accessories available separately.
Services: Caron provides customers with top-quality products backed by technical service and support for the life of the product. PM Service Kits help to minimize downtime by providing routine maintenance items for equipment. See table below for more information.
Documents
AT-Lit_7900-7901FreezeThawChambers
Hoja de datos
Especificaciones
- Volume:708 L (25 cu. ft.)
- Description:Freeze/thaw chamber, model 7900-25-2 with capacitive humidity sensor
- Interior Dimensions:81W×68.6D×133.9H cm (32×27×52.7")
- Electrical:208/230 V, 60 Hz,12 A
- Humidity Range:20 - 98% RH
- Shipping Weight:356 kg (785 lbs.)
- Cat. No.:10216-800
- Temperature uniformity:±0.3 °C
- Temperature sensor:3-wire RTD
- Temperature range:−25…+70 °C
- Temperature control:±0.1 °C
- Environmentally Preferable:Sustainable Packaging|Energy Efficient
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



