Orders LinkContactUsLinkComponent
Jar with Cap, HDPE
Jar with Cap, HDPE
Jar with Cap, HDPE, 1000 mL
Catalog # 470191-495
Supplier:   DYNALON
Número de proveedor #:  226415-1000
Jar with Cap, HDPE
Catalog # 470191-495
Supplier:  DYNALON
Jar with Cap, HDPE
Jar with Cap, HDPE
Jar with Cap, HDPE, 1000 mL
Price
$1,728.00
Log in for availability
Paquete de 10
Product Details & Documents
These cylindrical food-grade HDPE containers with black HDPE screw closures are supplied with a neck insert for added security, suitable for powders, solid specimens, creamy or semisolid substances, crystals, and hygroscopic substances.
  • Aligned with GLOBE protocols
Specifications
  • Description:
    Jar with Cap, HDPE, 1000 mL