$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
PLANTILLA CUADRADA PLST 4X5
35001-216
:
COPAN DIAGNOSTICS INC
PLANTILLA CUADRADA PLST 4X5
35001-216
:
COPAN DIAGNOSTICS INC
:
T2906CS01
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Disposable Petri Dishes
: VWR International