Streak Plates
Streak Plates, Black - 10/PK, 2x2"
Catalog # 470097-470
Supplier: SHIVDIAL SUD & SONS
Supplier #: B19182-SP
Jump to:
Product Details & Documents
Streak color helps identify minerals.
- Plates Sized for Field and Lab Testing
- Unglazed Porcelain
- Black Plates Work with Light Minerals
These unglazed porcelain plates are sized for field and lab testing. Rubbing the specimen across the plate reveals its streak color.
Black plates work with light minerals.
Specifications
- Description:Streak Plates, Black - 10/PK
- L×W:2x2"