Orders LinkContactUsLinkComponent
Microscope Slide Storage Boxes for 50 Slides, Globe Scientific
Microscope Slide Storage Boxes for 50 Slides, Globe Scientific
Orange
Catalog # 89510-812
Supplier #:  513077N
Microscope Slide Storage Boxes for 50 Slides, Globe Scientific
Catalog # 89510-812
Microscope Slide Storage Boxes for 50 Slides, Globe Scientific
Microscope Slide Storage Boxes for 50 Slides, Globe Scientific
Orange
Price
$138.40
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Globe's microscope slide storage boxes are designed for the safe and long term storage of microscope slides
An inventory sheet is located on the inside cover for cenvenient slide identification and organization. The base of the box is cork lined for added protection.
They are produced from durable ABS plastic and offer a hinged lid for easy opening and closing.
Ordering information: Available in 4 sizes, these storage boxes accommodate 25 x 75 mm and 3 x 1" microscope slides.
Especificaciones
  • Color:
    Orange