Microscope Slide Storage Boxes for 50 Slides, Globe Scientific
Dark Grey
Catalog # 89510-810
Supplier: GLOBE SCIENTIFIC INC
Número de proveedor #: 513077G
Microscope Slide Storage Boxes for 50 Slides, Globe Scientific
Catalog # 89510-810
Supplier: GLOBE SCIENTIFIC INC
Jump to:
Product Details & Documents
Globe's microscope slide storage boxes are designed for the safe and long term storage of microscope slides
An inventory sheet is located on the inside cover for cenvenient slide identification and organization. The base of the box is cork lined for added protection.
They are produced from durable ABS plastic and offer a hinged lid for easy opening and closing.
Ordering information: Available in 4 sizes, these storage boxes accommodate 25 x 75 mm and 3 x 1" microscope slides.
Documents
Globe Scientific Select Laboratory Supplies Specifically for Healthcare Brochure
Documentación del producto
Especificaciones
- Color:Dark Grey