Orders LinkContactUsLinkComponent
Biochemical Evidence for Evolution Lab Activity
Biochemical Evidence for Evolution Lab Activity, 10
Catalog # 470213-176
Supplier:   Avantor
Biochemical Evidence for Evolution Lab Activity
Catalog # 470213-176
Supplier:  Avantor
Biochemical Evidence for Evolution Lab Activity
Biochemical Evidence for Evolution Lab Activity
Biochemical Evidence for Evolution Lab Activity, 10
Price
$3,347.47
Log in Para disponibilidad
Each (1UN) Paleta de 45 (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Look at Evolution through the Study of Homology
  • Investigate evolutionary relationships with a modified Nuttall precipitation technique
  • Analyse similarities and differences between a variety of simulated animal blood sera
  • Materials included for ten lab setups
The study of evolution using homology consists of a classification method based on analysis of antigen-antibody complexes found in the blood. Using a modified Nuttall precipitation technique, students will identify the source of each sample. For safety, this activity uses five different simulated animal blood sera instead of the real thing. The Biochemical Evidence for Evolution Lab comes with enough materials for ten setups, a teacher’s guide, and student copymaster.
Especificaciones
  • Description:
    Biochemical Evidence for Evolution Lab Activity
  • No. of Lab Groups:
    10