Biochemical Evidence for Evolution Lab Activity
Product Details & Documents
Look at Evolution through the Study of Homology
- Investigate evolutionary relationships with a modified Nuttall precipitation technique
- Analyse similarities and differences between a variety of simulated animal blood sera
- Materials included for ten lab setups
The study of evolution using homology consists of a classification method based on analysis of antigen-antibody complexes found in the blood. Using a modified Nuttall precipitation technique, students will identify the source of each sample. For safety, this activity uses five different simulated animal blood sera instead of the real thing. The Biochemical Evidence for Evolution Lab comes with enough materials for ten setups, a teacher’s guide, and student copymaster.
Especificaciones
- Description:Biochemical Evidence for Evolution Lab Activity
- No. of Lab Groups:10
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.