Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polypropylene Volumetric Flasks with Screw Caps

Polypropylene Volumetric Flasks with Screw Caps

500 ml
Catalog # 470211-818
Número de proveedor #:  4000-0500
Polypropylene Volumetric Flasks with Screw Caps
Catalog # 470211-818
Polypropylene Volumetric Flasks with Screw Caps
Polypropylene Volumetric Flasks with Screw Caps

500 ml

Price
$1,950.00
Log in Para disponibilidad
Each (1UN) Case de 8 (1UN)
Product Details & Documents

Excellent Chemical Resistance

  • Calibrated “To Contain” and “To Deliver” at 20°C (68°F)
  • No meniscus to confuse readings
  • Threaded neck with polypropylene screw closure

Autoclavable (Remove closure before autoclaving).

Ordering information: Cap included but not shown.

Especificaciones
  • Capacity:
    500 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.