Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polypropylene Volumetric Flasks with Screw Caps

Polypropylene Volumetric Flasks with Screw Caps

50 ml
Catalog # 470211-812
Número de proveedor #:  4000-0050
Polypropylene Volumetric Flasks with Screw Caps
Catalog # 470211-812
Polypropylene Volumetric Flasks with Screw Caps
Polypropylene Volumetric Flasks with Screw Caps

50 ml

Price
$1,220.00
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Product Details & Documents

Excellent Chemical Resistance

  • Calibrated “To Contain” and “To Deliver” at 20°C (68°F)
  • No meniscus to confuse readings
  • Threaded neck with polypropylene screw closure

Autoclavable (Remove closure before autoclaving).

Ordering information: Cap included but not shown.

Especificaciones
  • Capacity:
    50 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.