Dropper Bottles
Dropper Bottles, 4 ml
Catalog # 470211-800
Supplier: Thermo Fisher Scientific
Número de proveedor #: 2752-9125
Jump to:
Product Details & Documents
These bottles are made of low-density polyethylene.
- Bottles Dispense about 40 mcL per Drop
- Good Chemical Resistance
- Semi-Transparent Bottles
- Polyethylene Construction
The semi-transparent bottles feature color-coded caps.
They have good chemical resistance and dispense about 40 mcL per drop.
Especificaciones
- Capacity:4 ml
- Description:Dropper Bottles