PYREX® Watch Glasses
Watch glass, 125 mm
Supplier: CORNING CA
Número de proveedor #: 9985-125
Jump to:
Product Details & Documents
PYREX® borosilicate glass.
- High chemical durability
- Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
Packaging: One package has 12 watch glasses.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Ø:125 mm
- Description:Watch glass