PYREX® Volumetric Pipettes, Color-Coded, Class A
Pyrex® transfer pipettes (volumetric pipettes), cap., each, 25 ml
Supplier: CORNING CA
Número de proveedor #: 7100-25
PYREX® Volumetric Pipettes, Color-Coded, Class A
Catalog # 470211-604
Supplier: CORNING CA
Jump to:
Product Details & Documents
These pipettes are made of high-grade borosilicate glass.
- Reusable, rugged construction
- Color-coded
- Colored enamel graduations and markings
Ruggedly constructed and properly proportioned, pipettes have accuracy tolerances to meet or exceed the strict requirements for Class A Volumetric Ware as indicated by ASTM E969.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:25 ml
- Description:Pyrex® transfer pipettes (volumetric pipettes), cap., each
- Cat. No.:470211-604
- Environmental certification:I-RECs, ISO 14001 - Monterrey
- Environmentally Preferable:Low Manufacturing Impact