Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Digital Hot Plates, Corning®

120 V, 60 Hz, PC-600D, 25...550 °C (77...1022 °F), 10×10"
Catalog # 470211-566
Supplier:   CORNING CA
 :  6795-600D
Digital Hot Plates, Corning®
Catalog # 470211-566
Supplier:  CORNING CA
Digital Hot Plates, Corning®
Digital Hot Plates, Corning®

120 V, 60 Hz, PC-600D, 25...550 °C (77...1022 °F), 10×10"

Price
$19,110.00
Log in Para disponibilidad
Each (1UN)
Product Details & Documents

High-quality, durable hot plates have added microprocessor technology with digital displays for optimal temperature control. All units are equipped with Pyroceram® glass-ceramic tops set atop rugged housing and heating elements. The digital LED temperature display is adjustable in 5°C increments, and blinks until set temperature is reached. Microprocessor control maintains consistent and repeatable settings.

  • Microprocessor maintains consistent and repeatable temperature settings from 5 °C (if ambient temperature is 0 °C or lower) up to 550 °C
  • Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
  • Optional external temperature controller (89090-224) truly controls temperature inside the vessel
  • Bright LED 'hot' icon lights up when top plate is over 60 °C, even when heat control is turned off
  • Two piece casting design deflects spills away from electronics and includes a built in mount for an optional 457 mm support rod
  • Two year manufacturer's warranty.

UL and C-UL listed. CE marked.

Corresponding magnetic hotplate stirrers (heating and stirring 89090-198 and 89090-216) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.

 : UL and C-UL listed. CE marked.

Especificaciones
  • Model:
    PC-600D
  • Temperature Range:
    25...550 °C (77...1022 °F)
  • Electrical:
    120 V, 60 Hz
  • Top Plate Dimensions:
    10×10"