Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

PYREX® Volumetric Flasks with [ST] Stopper

100 ml, Pyrex® Volumetric Flasks with Glass Stopper, 100 ml, 13, ±0.16 ml
Catalog # 470211-522
Supplier:   CORNING CA
 :  5641-100
PYREX® Volumetric Flasks with [ST] Stopper
Catalog # 470211-522
Supplier:  CORNING CA
PYREX® Volumetric Flasks with [ST] Stopper
PYREX® Volumetric Flasks with [ST] Stopper

100 ml, Pyrex® Volumetric Flasks with Glass Stopper, 100 ml, 13, ±0.16 ml

Price
$1,320.00
Log in Para disponibilidad
Each (1UN) Case de 12 (1UN)
Product Details & Documents

Made of PYREX® borosilicate glass.

  • Graduation line is sharp and permanent
  • Large white block letters make inscriptions easy to read
  • Supplied with interchangeable ground glass stoppers

Flasks are designed for those that do not require the precision accuracy of our Class A capacity flasks. Capacity tolerances are twice those specified for Class A volumetric ware.

Documents

Especificaciones
  • Capacity:
    100 ml
  • Description:
    Pyrex® Volumetric Flasks with Glass Stopper, 100 ml
  • ST Stopper No.:
    13
  • Tolerance:
    ±0.16 ml