PYREX® Filtering Flasks
500 ml
Supplier: CORNING CA
Número de proveedor #: 5340-500
Jump to:
Product Details & Documents
PYREX® borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks have a beaded top that provides an excellent fit for rubber stoppers. Graduated to show approximate volume.
The heavy-duty rim flange is ideal for supporting Buchner funnels. It includes a side arm for attaching rubber tubing. Extra-large marking spot for easy identifications.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Capacity:500 ml