Orders LinkContactUsLinkComponent
PYREX® Flat-Bottom Evaporating Dishes, Corning
PYREX® Flat-Bottom Evaporating Dishes, Corning
170 ml, 90×50 mm
Catalog # 470211-464
Supplier:   CORNING CA
Supplier #:  3180-90
PYREX® Flat-Bottom Evaporating Dishes, Corning
Catalog # 470211-464
Supplier:  CORNING CA
PYREX® Flat-Bottom Evaporating Dishes, Corning
PYREX® Flat-Bottom Evaporating Dishes, Corning
170 ml, 90×50 mm
Price
$607.00
Log in Para disponibilidad
Each (1UN) Case de 6 (1UN)
Product Details & Documents
PYREX® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Dishes with rugged beaded rims and standard medium depth.
Especificaciones
  • Capacity:
    170 ml
  • Diameter ×H:
    90×50 mm