PYREX® Student Burets with Replaceable Components
Pyrex® Replaceable Buret Tip, —, —
Supplier: CORNING CA
Número de proveedor #: 2116-TO
Jump to:
Product Details & Documents
Durable Enough for Repeated Student Use.
- Tooled tops and bottoms
- White enamel graduations
- Teflon® stopcock reduces breakage
The replaceable stopcock assemblies feature a screw thread locking nut and collar and a replaceable tip with ground end. Teflon® stopcock permits replacement of the body or tip. New triangular shipping package reduces breakage in transit. Replacement parts available.
Documents
Especificaciones
- Graduation Range:—
- Capacity:—
- Description:Pyrex® Replaceable Buret Tip