PYREX® Standard-Grade Beakers
800 ml, 50 ml, 50 - 750 ml
: 1000-800
Made of PYREX® borosilicate glass.
- PYREX®, Soda-lime glass, with spout
- Very good chemical resistance
- Good temperature resistance
Manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength.
Sizes 250 ml and larger have a double graduated metric scale to indicate approximate content. The 10 ml size is not graduated. All sizes have an extra large marking spot.
: Meet ASTM E960 specifications.
: The variety pack includes one of each of the following beakers: 50, 100, 250, 600 and 1000 ml.
- Graduation Interval:50 ml
- Graduation Range:50 - 750 ml
- Capacity:800 ml