Orders LinkContactUsLinkComponent
PYREX® Standard-Grade Beakers
800 ml, 50 ml, 50 - 750 ml
  470211-334
 :   CORNING CA
 :  1000-800
PYREX® Standard-Grade Beakers
  470211-334
PYREX® Standard-Grade Beakers
PYREX® Standard-Grade Beakers
800 ml, 50 ml, 50 - 750 ml
$1,890.00
Paquete de 6 Case de 4 (6UN)
Made of PYREX® borosilicate glass.
  • PYREX®, Soda-lime glass, with spout
  • Very good chemical resistance
  • Good temperature resistance
Manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength.
Sizes 250 ml and larger have a double graduated metric scale to indicate approximate content. The 10 ml size is not graduated. All sizes have an extra large marking spot.
 : Meet ASTM E960 specifications.
 : The variety pack includes one of each of the following beakers: 50, 100, 250, 600 and 1000 ml.
  • Graduation Interval:
    50 ml
  • Graduation Range:
    50 - 750 ml
  • Capacity:
    800 ml