Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

PYREX® Standard-Grade Beakers

10 ml (0.34 oz.), 1 ml, 1 - 10 ml
Catalog # 470211-308
Supplier:   CORNING CA
 :  1000-10
PYREX® Standard-Grade Beakers
Catalog # 470211-308
Supplier:  CORNING CA
PYREX® Standard-Grade Beakers
PYREX® Standard-Grade Beakers

10 ml (0.34 oz.), 1 ml, 1 - 10 ml

Price
$2,160.00
Log in Para disponibilidad
Paquete de 12 Case de 4 (12UN)
Product Details & Documents

Made of PYREX® borosilicate glass.

  • PYREX®, Soda-lime glass, with spout
  • Very good chemical resistance
  • Good temperature resistance

Manufactured with uniform wall thickness to offer optimum balance between thermal shock resistance and mechanical strength.

Sizes 250 ml and larger have a double graduated metric scale to indicate approximate content. The 10 ml size is not graduated. All sizes have an extra large marking spot.

 : Meet ASTM E960 specifications.

 : The variety pack includes one of each of the following beakers: 50, 100, 250, 600 and 1000 ml.

Especificaciones
  • Graduation Interval:
    1 ml
  • Graduation Range:
    1 - 10 ml
  • Capacity:
    10 ml (0.34 oz.)