Orders LinkContactUsLinkComponent
Culture Flasks
Culture Flasks
600 mL
Catalog # 470211-294
Supplier:   CORNING CA
Número de proveedor #:  430823
Culture Flasks
Catalog # 470211-294
Supplier:  CORNING CA
Culture Flasks
Culture Flasks
600 mL
Price
$708.00
Log in Para disponibilidad
Paquete de 5 Case de 10 (5UN)
Product Details & Documents
Specially Coated Tissue Culture Flasks.
  • Nunclon surface treatment promotes cell attachment and growth
  • Easy filling and access for pipets
  • Stackable, space saving design
Cell adherence is discouraged from the untreated sidewalls, neck and top panel.
Especificaciones
  • Description:
    600 mL