Culture Flasks
600 mL
Supplier: CORNING CA
Número de proveedor #: 430823
Jump to:
Product Details & Documents
Specially Coated Tissue Culture Flasks.
- Nunclon surface treatment promotes cell attachment and growth
- Easy filling and access for pipets
- Stackable, space saving design
Cell adherence is discouraged from the untreated sidewalls, neck and top panel.
Documents
Certificates
Buscar por número de loteThermo Scientific Nunc Cell Culture Plastics Guide
Documentación del producto
Especificaciones
- Description:600 mL