Balls
Foam ball, 3"
Catalog # 470211-258
Supplier: FLORA CRAFT CORPORATION
Número de proveedor #: BA3S/24
Jump to:
Product Details & Documents
Useful for collision and pendulum experiments.
A must have in any physics classroom or laboratory.
Especificaciones
- Description:Foam ball
- Diameter:3"